missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mitomycin C

Product Code. 30000846
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000846 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000846 Supplier R&D Systems™ Supplier No. 3258/2
  • ONLINE EXCLUSIVE PRICE
    138.12 €
    119.70 € / 2mg
    Save 18.42 €
    13% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. DNA cross-linking antitumor agent

Mitomycin C is an antibiotic and antitumor agent. Covalently binds DNA forming intra- and interstrand crosslinks. Inhibits DNA synthesis. Also used for MEF/STO feeder layer preparation in stem cell culture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-07-7
Molecular Formula C15H18N4O5

Specifications

Quantity 2 mg
Product Line Tocris
Formula Weight 334.33
Product Type Mitomycin C
Purity >98%
Molecular Weight (g/mol) 334.332
Chemical Name or Material Mitomycin C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.