missing translation for 'onlineSavingsMsg'
Learn More

Mitomycin C, Fisher BioReagents™

Product Code. 10182953
Change view
Click to view available options
Quantity:
2 mg
10 mg
20 mg
Packaging:
Amber Glass
Unit Size:
10mg
20mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10182953 2 mg Amber Glass 2mg
10022604 10 mg Amber Glass 10mg
10115004 20 mg Amber Glass 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10182953 Supplier Fisher BioReagents Supplier No. BP25312
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mitomycin C

Inhibitor of DNA synthesis, nuclear division, and cancer cells. Antibacterial spectrum: Gram positive, gram negative, acid-fast bacilli.

Recommended Storage:
4°C. Keep containers tightly closed in a dry, cool and well-ventilated place. Keep refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-07-7
Molecular Formula C15H18N4O5
Molecular Weight (g/mol) 334.332
InChI Key NWIBSHFKIJFRCO-WUDYKRTCSA-N
Synonym mitomycin c, mitomycin, ametycine, mutamycin, ametycin, mitocin-c, mitomycinum, mytomycin, mytozytrex, mitomycin-c
PubChem CID 5746
ChEBI CHEBI:27504
SMILES CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N

Specifications

Melting Point 360°C
Color Blue
Assay Percent Range ≥98.0 %
Packaging Amber Glass
Quantity 2 mg
Merck Index 15, 6300
Solubility Information Soluble in water
Formula Weight 334.3
Percent Purity 99.0% min. (RP CH3CN:H20 0 to 100%), 98.0% min. (RP MEOH:H20 90:10)
Physical Form Solid
Chemical Name or Material Mitomycin C
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Mitomycin C
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.