missing translation for 'onlineSavingsMsg'
Learn More

Mitotane-13C6, TRC

Product Code. 30478153
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478153 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478153 Supplier Toronto Research Chemicals Supplier No. TRCM3725031MG
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mitotane-13C6
CAS 1261396-21-7
Molecular Formula 13C6 C8 H10 Cl4
Quantity 1 mg
Synonym 1-Chloro-2-[2,2-dichloro-1-(4-chlorophenyl)ethyl]benzene-13C6, 1,1-Dichloro-2-(o-chlorophenyl)-2-(p-chlorophenyl)ethane-13C6, 1-(2-Chlorophenyl)-1-(4-chlorophenyl)-2,2-dichloroethane-13C6, 1-(2'-Chlorophenyl)-1-(4'-chlorophenyl)-2,2-dichloroethane-13C6, 1-(o-Chlorophenyl)-1-(p-chlorophenyl)-2,2-dichloroethane-13C6, 2,4'-Dichlorodiphenyldichloroethane-13C6, 2-(2-Chlorophenyl)-2-(4-chlorophenyl)-1,1-dichloroethane-13C6, 2-(p-Chlorophenyl)-2-(o-chlorophenyl)-1,1-dichloroethane-13C6, CB 313-13C6, Chloditan-13C6, Chlodithane-13C6, Lysodren-13C6, Mitotan-d4, NSC 38721-13C6, Opeprim-13C6, o,p'-DDD-13C6, o,p'-Dichlorodiphenyldichloroethane-13C6, o,p'-TD-13C6
InChI Formula InChI=1S/C14H10Cl4/c15-10-7-5-9(6-8-10)13(14(17)18)11-3-1-2-4-12(11)16/h1-8,13-14H/i5+1,6+1,7+1,8+1,9+1,10+1
SMILES ClC(Cl)C([13c]1[13cH][13cH][13c](Cl)[13cH][13cH]1)c2ccccc2Cl
IUPAC Name 1-chloro-2-[2,2-dichloro-1-(4-chloro(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl)ethyl]benzene
Molecular Weight (g/mol) 326
Formula Weight 323.9738
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.