missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MitoTracker™ Red CMXRos Dye, for flow cytometry

Product Code. 17521655
Change view
Click to view available options
Quantity:
5 vials x 20 tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17521655 5 vials x 20 tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17521655 Supplier Invitrogen™ Supplier No. M46752
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Mitotracker™ dyes for flow cytometry are cell permeant reagents that label mitochondria in live cells. Provided at an optimized concentration with protocols for flow cytometry to allow for more precise analysis with both conventional and spectral flow cytometry.

  • Cell permeant reagents that label mitochondria in live cells
  • Dyes are provided at an optimized concentration with protocols for flow cytometry to allows for more precise analysis with both conventional and spectral flow cytometry
  • Simply incubate cells in the recommended dye concentration for flow cytometry, which passively diffuses across the plasma membrane and accumulates in active mitochondria
  • Bright colors allow for easy detection of mitochondria using flow cytometry without the need for special filters
  • Dyes are consistent, reliable, and stable allowing for the generation of high-quality data
  • Optimized for flow cytometry use with all essential reagents for labeling mitochondria in a single kit
  • Dissolve a vial of dye in anhydrous DMSO to provide enough dye for 20 assays
  • A range of available colors are detected with standard flow cytometry filters
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Flow Cytometry
Format 5 x 20 tests
Label or Dye Dye
No. of Tests 100 tests
Quantity 5 vials x 20 tests
Regulatory Status RUO
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.