Learn More
Thermo Scientific Chemicals Mitoxantrone Dihydrochloride
Beskrivelse
Mitoxantrone dihydrochloride is an antiviral, antibacterial, antiprotozoal, immunomodulating, and antineoplastic cytostatic anthraquinone derivative. Induces DNA damage by intercalating into DNA and inhibiting Topo II (topoisomerase II). Mitoxantrone dihydrochloride induces interstrand DNA cross-links and DNA-protein cross-links in cellular systems. Mitoxantrone dihydrochloride has recently been shown to be an inhibitor of DNA methylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| Quantity | 50 mg |
| Description | Blue to black |
| Content And Storage | -30°C to -10°C |
| CAS | 70476-82-3 |
| Molecular Formula | C22H28N4O6.2HCl |
| Formula Weight | 517.40 |
| Synonym | 1,4-Dihydroxy-5,8-bis[[2-[(2-hydroxyethyl)amino]ethyl]amino]-9,10-anthracenedione dihydrochloride; |
| Solubility | Soluble to 5 mM in water and to 75 mM in DMSO |
| Product Type | Mitoxantrone dihydrochloride |
| Form | Crystalline solid |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.