Learn More
SP Genevac™ miVac™ 2 Stacker for swing rotor
Product Code. 31723011
Change view
Click to view available options
Type:
Adapter Set
Unit Size:
Each
Product Code. Type Unit Size
31723011 Adapter Set Each
1 options
Product Code. 31723011 Supplier SP Genevac™ Supplier No. BRSSTK00016
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Allows user to stack up to 5 shallow well plate per swing.
  • This Stacker accessory for miVac allows for up to 5 shallow well plate per swing.
Specifications
Type Adapter Set