Learn More
SP Genevac™ miVac™ DNA and Duo Rotor for glass vials
Product Code. 31723034
Change view
Click to view available options
Length (Metric):
40 mm
60 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
31723034 40 mm Each
31723035 60 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31723034 Supplier SP Genevac™ Supplier No. DRC18V40048
Only null left
Add to basket
Add to basket
Designed for efficient processing of multiple glass vials.
  • Manufactured from solid aluminum
  • Fits vials
Specifications
Type Rotor
Holds 48 Vials
Length (Metric) 40 mm
Diameter (Metric) 18 mm