Learn More
SP Genevac™ miVac™ Quattro Rotor for Dionex ASE™ 60 mL Vials
Product Code. 17831508
Change view
Click to view available options
Holds:
60 mL Vials
Product Code. Holds
17831508 60 mL Vials
1 options
Product Code. 17831508 Supplier SP Genevac™ Supplier No. QRC28V140030
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Solid Aluminum rotor can hold up to 30 - 60 mL Dionex ASE™ vials.
  • Manufactured from solid aluminum
  • Fits 60 mL Dionex ASE™ vials
  • Can hold up to 30 vials
Specifications
Type Rotor
Holds 60 mL Vials