Learn More
SP Genevac™ miVac™ Replacement lid seal
Product Code. 31723036
Change view
Click to view available options
For Use With (Equipment):
Duo Concentrator
Quattro Concentrator
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31723036 Quattro Concentrator Each
18897304 Duo Concentrator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31723036 Supplier SP Genevac™ Supplier No. 600002
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Replacement Seal for miVac Concentrator
  • Designed as a direct replacement lid seal for miVac concentrators to maintain optimal sealing performance
  • Compatible with both miVac Duo and Quattro Concentrator systems for versatile use
  • Helps ensure proper vacuum conditions for efficient sample concentration
  • Manufactured for reliable fit and long-lasting performance in laboratory environments
Specifications
Material EPDM
Type Seal
For Use With (Equipment) Quattro Concentrator