missing translation for 'onlineSavingsMsg'
Learn More

Mivacurium Chloride (mixture of isomers), TRC

Product Code. 30133865
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133865 2.5 mg 2.5mg
30010700 10 mg 10mg
30004889 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133865 Supplier Toronto Research Chemicals Supplier No. TRCM3850002.5MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106861-44-3
Molecular Formula C58 H80 N2 O14 . 2 Cl
Molecular Weight (g/mol) 1100.17
Synonym Isoquinolinium, 2,2'-[[(4E)-1,8-dioxo-4-octene-1,8-diyl]bis(oxy-3,1-propanediyl)]bis[1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-1-[(3,4,5-trimethoxyphenyl)methyl]-, chloride (1:2), (1R,1'R)-, Isoquinolinium, 2,2'-[(1,8-dioxo-4-octene-1,8-diyl)bis(oxy-3,1-propanediyl)]bis[1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-1-[(3,4,5-trimethoxyphenyl)methyl]-, dichloride, [R-[R*,R*-(E)]]-, Isoquinolinium, 2,2'-[[(4E)-1,8-dioxo-4-octene-1,8-diyl]bis(oxy-3,1-propanediyl)]bis[1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-1-[(3,4,5-trimethoxyphenyl)methyl]-, dichloride, (1R,1'R)- (9CI), BW B1090U dichloride, BW-B 1090U, Mivacron, Mivacurium chloride, [R-[R*,R*-(E)]]-2,2'-[(1,8-Dioxo-4-octene-1,8-diyl)bis(oxy-3,1-propanediyl)]bis[1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-1-[(3,4,5-trimethoxyphenyl)methyl]isoquinolinium dichloride
IUPAC Name bis[3-[(1R)-6,7-dimethoxy-2-methyl-1-[(3,4,5-trimethoxyphenyl)methyl]-3,4-dihydro-1H-isoquinolin-2-ium-2-yl]propyl] (E)-oct-4-enedioate;dichloride
SMILES [Cl-].[Cl-].COc1cc2CC[N+](C)(CCCOC(=O)CC\C=C\CCC(=O)OCCC[N+]3(C)CCc4cc(OC)c(OC)cc4[C@H]3Cc5cc(OC)c(OC)c(OC)c5)[C@H](Cc6cc(OC)c(OC)c(OC)c6)c2cc1OC

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C58H80N2O14.2ClH/c1-59(25-21-41-35-47(63-3)49(65-5)37-43(41)45(59)29-39-31-51(67-7)57(71-11)52(32-39)68-8)23-17-27-73-55(61)19-15-13-14-16-20-56(62)74-28-18-24-60(2)26-22-42-36-48(64-4)50(66-6)38-44(42)46(60)30-40-33-53(69-9)58(72-12)54(34-40)70-10;;/h13-14,31-38,45-46H,15-30H2,1-12H3;2*1H/q+2;;/p-2/b14-13+;;/t45-,46-,59?,60?;;/m1../s1
Formula Weight 1098.4987
Chemical Name or Material rac Mivacurium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.