missing translation for 'onlineSavingsMsg'
Learn More

MK 1775, TRC

Product Code. 30179955
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179955 50 mg 50mg
30179934 5 mg 5mg
30179804 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179955 Supplier Toronto Research Chemicals Supplier No. TRCM42497550MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 955365-80-7
Molecular Formula C27H32N8O2
Molecular Weight (g/mol) 500.6
Synonym Adavosertib, MK-1775, MK 1775
SMILES CN1CCN(CC1)c2ccc(Nc3ncc4C(=O)N(CC=C)N(c5cccc(n5)C(C)(C)O)c4n3)cc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C27H32N8O2/c1-5-13-34-25(36)21-18-28-26(29-19-9-11-20(12-10-19)33-16-14-32(4)15-17-33)31-24(21)35(34)23-8-6-7-22(30-23)27(2,3)37/h5-12,18,37H,1,13-17H2,2-4H3,(H,28,29,31)
Chemical Name or Material 1,2-Dihydro-1-[6-(1-hydroxy-1-methylethyl)-2-pyridinyl]-6-[(4-(4-methyl-1-piperazinyl)phenyl)amino]-2-(2-propen-1-yl)-3H-pyrazolo[3,4-d]pyrimidin-3-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.