missing translation for 'onlineSavingsMsg'
Learn More

MK-886, TRC

Product Code. 30478488
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478488 5 mg 5mg
30469717 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478488 Supplier Toronto Research Chemicals Supplier No. TRCM4250535MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118414-82-7
Molecular Formula C27H34ClNO2S
Molecular Weight (g/mol) 472.08
Synonym MK-886, L 663536
SMILES CC(C)c1ccc2c(c1)c(SC(C)(C)C)c(CC(C)(C)C(=O)O)n2Cc3ccc(Cl)cc3

Specifications

Quantity 5 mg
Chemical Name or Material 1-[(4-Chlorophenyl)methyl]-3-[(1,1-dimethylethyl)thio]-alpha,alpha-dimethyl-5-(1-methylethyl)-1H-indole-2-propanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.