missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (±)-ML 209
GREENER_CHOICE

Product Code. p-7112413
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18791068

Brand: Tocris Bioscience™ 5987/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

RORγt inverse agonist; suppresses Th17 cell differentiation

(±)-ML 209 is a RORγt inverse agonist (IC50 = 460 nM). Suppresses Th17 cell differentiation and IL17A expression in vitro. Does not suppress transcription of RORα. Minimal activity displayed on ERα, LXRα and thyroid hormone receptors (IC50 values >4.4 μM).
TRUSTED_SUSTAINABILITY

Specifications

CAS 1334526-14-5
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Target Nuclear Receptor Antagonists
Molecular Formula C25H31NO6
Formula Weight 441g/mol
Color White
Product Type Inhibitor

For Laboratory Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.