missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 233
GREENER_CHOICE

Product Code. 18754828
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18754828

Brand: Tocris Bioscience™ 5966/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Non-peptide apelin receptor agonist

ML 233 is a non-peptide apelin receptor agonist (EC50 = 3.7 μM); exhibits >21-fold selectivity over the closely related angiotensin 1 receptor (EC50 = >79 μM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-4-Methyl-5-(((phenylsulfonyl)oxy)imino)-[1,1'-bi(cyclohexane)]-3,6-dien-2-one
CAS 143621-35-6
Quantity 10 mg
Solubility Soluble to 50mM in DMSO
Molecular Formula C19H21NO4S
Formula Weight 359.44
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.