missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 277

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ML 277 is a selective Kv7.1 (KCNQ1) potassium channel activator (EC50 = 260 nM). Exhibits >100-fold selectivity versus KCNQ2, KCNQ4 and hERG potassium channels. Augments IKs current of cultured human cardiomyocytes and shortens action potential duration.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | ML 277 |
| Purity | >98% |
| Molecular Weight (g/mol) | 471.59 |
| Inhibitors | ML 277 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction