missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 277
GREENER_CHOICE

Product Code. p-5545888
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18721608

Brand: Tocris Bioscience™ 4777/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective Kv7.1 (KCNQ1) potassium channel activator; augments IKs current

ML 277 is a selective Kv7.1 (KCNQ1) potassium channel activator (EC50 = 260 nM). Exhibits >100-fold selectivity versus KCNQ2, KCNQ4 and hERG potassium channels. Augments IKs current of cultured human cardiomyocytes and shortens action potential duration.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type ML 277
Purity >98%
Molecular Weight (g/mol) 471.59
Inhibitors ML 277

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.