missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 323

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
ML 323 is a potent and selective allosteric inhibitor of USP1-UAF1 deubiquitinase complex (IC50 = 76 nM). Exhibits selectivity for USP1-UAF1 over a panel of enzymes, including other deuquitinating enzymes. Inhibits deubiquitination of PCNA and FANCD2 and sensitizes cisplatin-resistant H596 cells to cell death induced by cisplatin (Cat. No. 2251) in vitro.
Specifications
Specifications
| CAS | 1572414-83-5 |
| Synonym | 5-Methyl-2-[2-(1-methylethyl)phenyl]-N-[[4-(1H-1,2,3-triazol-1-yl)phenyl]methyl]-4-pyrimidinamine |
| Quantity | 5 mg |
| Molecular Formula | C23H24N6 |
| Formula Weight | 384.48 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction