Learn More
Tocris Bioscience™ ML 349
Selective and reversible lysophospholipase 2 (LYPLA2) inhibitor
Brand:  Tocris Bioscience™ 5344/50
Encompass
Product Code. 15746736
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
ML 349 is a selective and reversible lysophospholipase 2 (LYPLA2) inhibitor (IC50 = 144 nM). Displays selectively over LYPLA1 and 20 other serine hydrolases (IC50 values are >3000 and >10,000 nM respectively).
Specifications
Specifications
>98%