Learn More
Tocris Bioscience™ ML 382
GREENER_CHOICE
MRGPRX1 positive allosteric modulator
Brand:  Tocris Bioscience™ 5738/10
Encompass
Product Code. 15737086
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
ML 382 is a selective MRGPRX1 positive allosteric modulator (EC50 = 190 nM). Exhibits selectivity for MrgX1 over MrgX2, MrgC11 and a panel of 68 other GPCRs, ion channels and transporters. Enhances BAM (8-22) potency at MRGPRX1 by 7-fold.
Specifications
Specifications
Mas-Related G Protein Coupled Receptor Modulators
1646499-97-9