missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 67-33

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ML 67-33 is a K2P potassium channel activator (EC50 values are 21.8 - 29.4 μM, 30.2 μM and 27.3 μM for K2P2.1 (TREK-1), K2P10.1 (TREK-2) and K2P4.1 (TRAAK) respectively, expressed in xenopus oocytes). Increases channel currents by activating core gating apparatus of channels.
Specifications
Specifications
| Quantity | 50 mg |
| Molecular Formula | C18H17Cl2N5 |
| Formula Weight | 374.27 |
| Product Type | Channel Activator |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction