missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 67-33

Description
ML 67-33 is a K2P potassium channel activator (EC50 values are 21.8 - 29.4 μM, 30.2 μM and 27.3 μM for K2P2.1 (TREK-1), K2P10.1 (TREK-2) and K2P4.1 (TRAAK) respectively, expressed in xenopus oocytes). Increases channel currents by activating core gating apparatus of channels.
Specifications
Specifications
| Quantity | 10 mg |
| Molecular Formula | C18H17Cl2N5 |
| Formula Weight | 374.27 |
| Product Type | Channel Activator |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?