missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 67-33
GREENER_CHOICE

Product Code. p-7166825
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30000782

Brand: Tocris Bioscience™ 6886/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

K2P2.1 (TREK-1), K2P10.1 (TREK-2) and K2P4.1 (TRAAK) channel activator

ML 67-33 is a K2P potassium channel activator (EC50 values are 21.8 - 29.4 μM, 30.2 μM and 27.3 μM for K2P2.1 (TREK-1), K2P10.1 (TREK-2) and K2P4.1 (TRAAK) respectively, expressed in xenopus oocytes). Increases channel currents by activating core gating apparatus of channels.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Molecular Formula C18H17Cl2N5
Formula Weight 374.27
Product Type Channel Activator
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.