missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies MLA™ Brand Macro Pipetters
For large volume handling tasks such as reconstituting reagents, controls, or dilutions
Brand: VistaLab™ Technologies 1070
Additional Details : Weight : 0.50000kg
Description
- Factory calibrated for accurate volume delivery
- Can be easily adjusted to achieve accurate performance on dense or viscous liquids
Specifications
±0.6% | |
Aluminum, Stainless Steel | |
Macropipette | |
Nozzle Insert, Calibration Key | |
Lifetime | |
2.5 in. |
Red | |
1 | |
10 mL | |
1 Ea. | |
11 in. | |
1.5 in. |