missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies MLA™ Brand Macro Pipetters
For large volume handling tasks such as reconstituting reagents, controls, or dilutions
399.00 € - 525.00 €
Specifications
Material | Aluminum, Stainless Steel |
---|---|
Number of Channels | 1 |
Type | Macropipette |
Includes | Nozzle Insert, Calibration Key |
Quantity | 1 Ea. |
Product Code | Brand | Accuracy | Color | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Accuracy | Color | Volume (Metric) | Price | Quantity & Availability | |||
11905842
|
VistaLab™ Technologies
1077 |
±0.6% | Red | 7 mL |
525.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
11945832
|
VistaLab™ Technologies
1067 |
±0.75% | Green | 3 mL |
400.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
11975832
|
VistaLab™ Technologies
1070 |
±0.6% | Red | 10 mL |
515.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
11965832
|
VistaLab™ Technologies
1069 |
±0.75% | Yellow | 5 mL |
399.00 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
11955832
|
VistaLab™ Technologies
1068 |
±0.75% | White | 4 mL |
400.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
11935832
|
VistaLab™ Technologies
1066 |
±0.75% | Red | 2 mL |
400.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
11905822
|
VistaLab™ Technologies
1065 |
±0.75% | Gray | 1 mL |
400.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
11915842
|
VistaLab™ Technologies
1078 |
±0.6% | Red | 8 mL |
515.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
11925842
|
VistaLab™ Technologies
1079 |
±0.6% | Red | 9 mL |
515.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
11995832
|
VistaLab™ Technologies
1076 |
±0.6% | Red | 6 mL |
515.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
Description
- Factory calibrated for accurate volume delivery
- Can be easily adjusted to achieve accurate performance on dense or viscous liquids
Specifications
Aluminum, Stainless Steel | |
Macropipette | |
1 Ea. | |
11 in. | |
1.5 in. |
1 | |
Nozzle Insert, Calibration Key | |
Lifetime | |
2.5 in. |