Learn More
VistaLab™ Technologies MLA™ D-Tipper™ Pipetters
Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts). Color coded body for easy volume identification and individually serialized with Calibration Certificate included.
322.00 € - 1630.00 €
Specifications
Autoclavable | Not Autoclavable |
---|---|
Model | D-Tipper™ |
Number of Channels | 1 |
Sterility | Non-sterile |
Type | Axial Pipette |
Product Code | Brand | Accuracy | Volume (Metric) | Precision | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Accuracy | Volume (Metric) | Precision | Price | Quantity & Availability | |||
11985882
|
VistaLab™ Technologies
1159C |
±1% | 600 μL | 1.00% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11995892
|
VistaLab™ Technologies
1162C |
±1% | 700 μL | 0.86% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11975882
|
VistaLab™ Technologies
1157C |
±1% | 700 μL | 0.80% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11925862
|
VistaLab™ Technologies
1142C |
±5% | 15 μL | 1.73% |
324.00 €
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||
11945862
|
VistaLab™ Technologies
1144C |
±3% | 40 μL | 0.70% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11965862
|
VistaLab™ Technologies
1146C |
±2% | 60 μL | 0.67% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11975862
|
VistaLab™ Technologies
1147C |
±2% | 70 μL | 0.57% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11985862
|
VistaLab™ Technologies
1148C |
±2% | 75 μL | 0.53% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11995862
|
VistaLab™ Technologies
1149C |
±2% | 80 μL | 0.50% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11905872
|
VistaLab™ Technologies
1150C |
±2% | 90 μL | 0.44% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11915872
|
VistaLab™ Technologies
1151C |
±1% | 120 μL | 0.45% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11925872
|
VistaLab™ Technologies
1152C |
±1% | 150 μL | 0.36% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11985892
|
VistaLab™ Technologies
1163C |
±1% | 900 μL | 0.67% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11935862
|
VistaLab™ Technologies
1143C |
±3% | 30 μL | 0.93% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11935872
|
VistaLab™ Technologies
1153C |
±1% | 400 μL | 0.50% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11955792
|
VistaLab™ Technologies
1053C |
±3% | 25 μL | 1.12% |
322.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11995792
|
VistaLab™ Technologies
1056C |
±1% | 200 μL | 0.30% |
322.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11975802
|
VistaLab™ Technologies
1057C |
±1% | 250 μL | 0.72% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11985802
|
VistaLab™ Technologies
1058C |
±1% | 300 μL | 0.60% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11935792
|
VistaLab™ Technologies
1051C |
±5% | 10 μL | 2.60% |
322.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11945792
|
VistaLab™ Technologies
1052C |
±3% | 20 μL | 1.35% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11965792
|
VistaLab™ Technologies
1054C |
±2% | 50 μL | 0.80% |
322.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11975792
|
VistaLab™ Technologies
1055C |
±1% | 100 μL | 0.54% |
322.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11995802
|
VistaLab™ Technologies
1059C |
±1% | 500 μL | 0.40% |
324.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11985812
|
VistaLab™ Technologies
1060C |
±1% | 1000 μL | 0.60% |
322.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
11915862
|
VistaLab™ Technologies
1141C |
±5% | 5 μL | 3.74% |
324.00 €
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||
11935942
|
VistaLab™ Technologies
1050-0999 |
±5.00%/ ±2.00%/ ±1.00%/ ±1.00%/ ±1.00% | 10, 50, 100, 200, 1000 μL | 2.60%/0.80%/0.54%/0.30%/0.60% |
1630.00 €
1 set |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
Description
- Single-stroke “to deliver” operation
- Built-in tip-ejector and glove guard
- Factory calibrated to deliver specific quantities; can be easily recalibrated in the lab
- Available in 30 fixed volumes ranging from 5-1000 μL
- Also available as a Selectable model: 50/100/200 μL
Specifications
Not Autoclavable | |
1 | |
Axial Pipette | |
Limited Lifetime |
D-Tipper™ | |
Non-sterile | |
Micro Pipetting/Liquid Transfer |