missing translation for 'onlineSavingsMsg'
Learn More

MLN120B, MedChemExpress

Product Code. 16472647
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16472647

missing translation for 'mfr': Medchem Express HY15473

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MLN120B (ML120B) is a potent, ATP competitive, and orally active inhibitor of IKKβ with an IC50 of 60 nM. MLN120B inhibits multiple myeloma cell growth in vitro and in vivo and also can be used for the research of rheumatoid arthritis.

MLN120B (ML120B) is a potent, ATP competitive, and orally active inhibitor of IKKβ with an IC50 of 60 nM. MLN120B inhibits multiple myeloma cell growth in vitro and in vivo and also can be used for the research of rheumatoid arthritis.

Specifications

Chemical Name or Material MLN120B
CAS 783348-36-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H15ClN4O2
Quantity 50 mg
Synonym ML120B
Solubility Information DMSO : ≥ 31 mg/mL (84.51 mM)
SMILES O=C(C1=CC=CN=C1C)NC2=C(OC)C(Cl)=CC3=C2NC4=C3C=CN=C4
Molecular Weight (g/mol) 366.8
Formula Weight 366.8
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.