missing translation for 'onlineSavingsMsg'
Learn More

MMAF-OMe, MedChemExpress

Product Code. 16488238
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488238 10 mg 10mg
16460688 5 mg 5mg
16420366 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488238 Supplier Medchem Express Supplier No. HY79256
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress MMAF-Ome, an antitubulin agent, is also an ADC cytotoxin. MMAF-Ome inhibits several tumor cell lines with IC50s of 0.056 nM, 0.166 nM, 0.183 nM, and 0.449 nM for MDAMB435/5T4, MDAMB361DYT2, MDAMB468, and Raji (5T4-) cell lines, respectively.

MMAF-Ome, an antitubulin agent, is also an ADC cytotoxin. MMAF-Ome inhibits several tumor cell lines with IC50s of 0.056 nM, 0.166 nM, 0.183 nM, and 0.449 nM for MDAMB435/5T4, MDAMB361DYT2, MDAMB468, and Raji (5T4-) cell lines, respectively.

Specifications

Chemical Name or Material MMAF-OMe
CAS 863971-12-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C40H67N5O8
Quantity 10 mg
Synonym Monomethyl auristatin F methyl ester
Solubility Information DMSO : ≥ 100 mg/mL (134.05 mM)
SMILES CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C(OC)=O)CC2=CC=CC=C2)=O)OC)=O)OC)N(C([C@H](C(C)C)NC([C@H](C(C)C)NC)=O)=O)C
Molecular Weight (g/mol) 745.99
Formula Weight 745.99
Percent Purity 96.68%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.