missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mMESSAGE mMACHINE™ T7 mRNA Kit with CleanCap™ Reagent AG

Product Code. 17904741
Change view
Click to view available options
No. of Reactions:
1000 Reactions
25 Reactions
50 Reactions
Unit Size:
1000 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
17904741 50 Reactions 50 reactions
31662617 25 Reactions -
17994731 1000 Reactions 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17904741 Supplier Invitrogen™ Supplier No. A57620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG contains all the reagents needed to generate mRNA with naturally occurring Cap 1 structure while providing increased yields and higher quality with greater translatability in vivo compared to legacy capping methods such as ARCA caps.

The mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG contains all the reagents and buffers necessary for high quality in vitro synthesis of large amounts of mRNA with the naturally occurring Cap 1 structure. Utilization of the CleanCap Reagent AG ensures superior results relative to older capping technologies and greater protein production. Each standard 20-μL reaction yields a minimum of 100 μg of Cap 1-containing mRNA, while the scaled-up protocol yields up to 1 mg of mRNA. This kit is suitable for full- or partially-modified-nucleotide substitution to generate modified mRNA.

Features of the mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG include:

  • Contains proven CleanCap capping technology
  • Optimized to achieve high mRNA yields (>5 mg/mL)
  • Produces mRNA that is over 95% capped
  • Optimized for production of mRNA from 0.9–10 kb templates
  • Minimum yields of 100 μg per reaction, up to 150 μg per reaction (see figure below)
  • Allows use of modified nucleotides, full- or partial-modified substitution
  • Includes linearized 0.9 kb GFP control template, yields ≥100 μg mRNA
  • Made up of high quality TheraPure reagents to enable transition from RUO to GMP
  • Available in two sizes: 50 and 1000 reactions*

*Note: 1000 reaction kit is designed to support scale-up to large amounts of mRNA production enabling ≥100 mg mRNA.

The mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG contains the trinucleotide cap analog CleanCap Reagent AG, which has the Cap 1 structure and can be used for co-transcriptional capping of mRNA without compromising yield. This reagent produces high capping efficiencies compared to legacy capping reagents such as ARCA. Cap 1-containing mRNAs are considerably more stable and have superior translational activities compared to Cap 0 mRNAs produced using ARCA or mCap cap analogs.

A linearized DNA template containing an AG-initiating T7 RNA polymerase promoter is strongly recommended for this kit to achieve mRNA yields >5 mg/mL with over 90% of the transcripts capped. The mRNA produced with this kit can be used for many applications, including transfections, preclinical mRNA therapeutic mRNA studies, preclinical vaccine development, microinjections, in vitro translation, and RNA structure and function analysis.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type HEK293, 293, HeLa, CHO-K1, CHO, Jurkat, Primary Human T Cells, Primary Cells, Stem Cells, iPSC, NIH-3T3, HepG2, A549, HUVEC
Final Product Type Synthetic mRNA, mRNA
Format Animal Origin Free Kit
Isolation Technology Silica Spin Column
Purification Target mRNA, Synthetic mRNA
Sample Type mRNA, Synthetic mRNA
No. of Reactions 50 Reactions
Product Line mMESSAGE mMACHINE
Product Type T7 In Vitro Transcription kit
Quantity 50 Reactions
Promoter Modified T7
Shipping Condition Dry Ice
Yield >100 μg
Starting Material PCR Products, Linearized DNA Plasmid
Show More Show Less
How is CleanCap Reagent AG different from ARCA (Anti-Reverse Cap Analog) technology?

CleanCap is a new capping technology that can provide higher mRNA yields and capping efficiencies than ARCA. CleanCap contains the cap 1 structure, which is found in higher eukaryotes such as mice or humans. ARCA contains the cap 0 structure that is found in lower eukaryotes such as yeast. Cap 1 mRNA can have superior translational activities in humans compared to Cap 0 mRNAs.

How can I introduce the RNA synthesized using the mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG (Cat. No. A57620, A57621), into cells?

The RNA can be introduced into cells by electroporation or transfection. Our Neon NxT Electroporation System can be used to introduce the RNA by electroporation. Our Lipofectamine MessengerMAX Transfection Reagent is designed for mRNA delivery and works with many cell types.

How can I purify the RNA synthesized using the mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG (Cat. No. A57620, A57621)?

The RNA synthesized using the mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG (Cat. No. A57620, A57621) can be purified using the provided LiCl Precipitation Solution. We recommend referring to the user guide for details. Alternatively, the RNA can be purified using spin columns such with the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908). Note that the MEGAclear Transcription Clean-Up Kit can purify up to 500 µg RNA and that it only purifies RNAs >100 nucleotides. If shorter RNAs need to be purified, we recommend using the GeneJET RNA Cleanup and Concentration Micro Kit (Cat. No. K0841).

Can I use modified nucleotides with the mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG (Cat. No. A57620, A57621)?

Yes, modified nucleotides can be used. We recommend referring to the user guide for guidance on this.

With the mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG (Cat. No. A57620, A57621), do you recommend using a template encoded poly-A tail?

Yes, we recommend using a template encoded poly-A tail as it yields better expression than when a poly-A tail is added enzymatically using poly-A polymerase. Refer to the user guide for guidance on making a template with a poly-A tail.

With the mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG (Cat. No. A57620, A57621), do you recommend using a linearized plasmid or a PCR product as the template?

Both types of templates are suitable for making mRNA with these kits and it is a matter of preference as to which one is used. We find that it is easier to get the amounts of template needed for large scale reactions (e.g., 1 mg mRNA or more) when using linearized plasmid as the template.

Do I need to change my promoter sequence to use CleanCap Reagent AG?

Yes, the promoter sequence should be modified. CleanCap Reagent AG needs an AG initiating promoter to achieve high capping efficiencies (>90%). Refer to the user guide for guidance on how to design the promoter so that it is compatible with CleanCap Reagent AG.

Can I handle CleanCap mRNA the same way as ARCA (Anti-Reverse Cap Analog) mRNA?

CleanCap mRNA can be handled the same way as ARCA mRNA.


For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.