missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MMP-9 Human, Biotrak assay

Product Code. 10197434 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
Each
Unit Size:
96 reactions
Product Code. Quantity unitSize
10197434 Each 96 reactions
1 options
This item is not returnable. View return policy

Product Code. 10197434

Brand: Cytiva RPN2614

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Anti-MMP-9 Microplate; Assay buffer; Standard; Peroxidase conjugate; Wash buffer; TMB substrate

Provides a simple, specific and precise quantitative determination of MMP-9 in tissue culture supernatants and homogenates as well as plasma

  • Specific for free pro-MMP-9 and pro-MMP-9 complexed to TIMP-1
  • No cross-reactivity with pro-MMP-1, -2, -3, TIMP-1 and -2
  • Sufficient material for 96 wells. If one standard curve is constructed, 40 unknowns can be measured in duplicate.
  • Range: 1-32ng/mL (4–128 ng/ml with plasma)
  • Sensitivity: 0.6ng/mL
  • Four-hour protocol

TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-9
Assay Range 1 to 32ng/mL
Cross Reactivity Human
Description MMP-9, Human, Biotrak Assay.
For Use With (Equipment) Pipettes or pipetting equipment with disposable polypropylene tips (100 μl, 500 μl, 1 mL and 5 mL); Disposable polypropylene test tubes; Glass measuring cylinders (100 mL and 500 mL); Distilled or deionized water; Spectrophtometric plate reader capable of
Includes Microplate (12 x 8-well strip coated with Anti-MMP-9), 10mL Phosphate Assay Buffer, 64ng Human pro MMP-9 standard(lyophilized), Anti-MMP-9 HRP Conjugate (lyophilized), 12.5mL Wash Buffer, TMB Substrate
Quantity Each
Sample Type Serum, Plasma (EDTA, Heparin, Citrate), Cell Culture Supernatant
Sensitivity 0.6 ng/mL
Total Assay Time 4 hr.
Content And Storage -15°C to -30°C
For Use With (Application) Suitable for use with serum, plasma (EDTA, heparin, citrate) and cell culture supernatant samplesBiotrak ELISAs measure MMP protein levels in human serum, plasma, and cell culture samples
Material Polypropylene
Product Type Human MMP-9 activity assay
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.