missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ MN Positive Pressure Frame

Product Code. 16311616
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16311616 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16311616 Supplier Macherey-Nagel™ Supplier No. 740474
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Adaptor for direct filtration using NucleoSpin™ 96 Plasmid kits on the [MPE]² positive pressure module

  • Adaptation of the NucleoSpin™ 96 Plasmid Transfection-grade kit procedure for the [MPE]² positive pressure module to speed up your extraction workflow
  • Reliable performance and excellent yields using NucleoSpin™ 96 Plasmid Transfection-grade on the [MPE]² positive pressure module
  • Low endotoxin levels comparable to standard vacuum processing
  • Compact and automated plasmid isolation of 96 samples in 60–90 minutes

Specifications

Format Adapter Frame
For Use With (Application) Plasmid DNA Purification
For Use With (Equipment) Hamilton [MPE]² Unit
Isolation Technology NucleoSpin Bead Tubes
Quantity 1 Ea.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.