Learn More
asecos™ Mobile Box Sorbents Refill Kit
Product Code. 17876486 Shop All asecos Products
Change view
Click to view available options
Type:
Chemical Spill Control Absorbent Storage Cart Refill Kit
Oil Spill Control Absorbent Storage Cart Refill Kit
Universal All Purpose Absorbent Storage Cart Refill Kit
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
17876486 Universal All Purpose Absorbent Storage Cart Refill Kit Each
17816546 Oil Spill Control Absorbent Storage Cart Refill Kit Each
17866546 Chemical Spill Control Absorbent Storage Cart Refill Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17876486 Supplier asecos™ Supplier No. 14409
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Mobile Box Sorbents Refill Kit is a practical refill set for mobile spill kit.
  • content: 6 cushions, 3 socks, 20 mats
Specifications
Material Heavy-duty Cloth
Type Universal All Purpose Absorbent Storage Cart Refill Kit
Weight (English) 6.61 lb.
Weight (Metric) 3 kg