Learn More
Miele Professional™ Mobile Injector
Shop All Miele Products
missing translation for 'orderingAttributeHoverText'
:
Includes: 37 Injector Nozzles
Description
- Equipped with 37 injector nozzles and locking mechanisms
- Equipped with connection for hot air drying
- Bracket for ML/2 for automatic mobile unit recognition
Specifications
Specifications
| Product Type | Mobile Injector Unit 1/1 |
| Model | E 414 |
| For Use With (Equipment) | Narrow-Necked Glassware |
| Includes | 37 Injector Nozzles |
| Quantity | 1 Each |