Learn More
Miele Professional™ Mobile Injector
Mobile unit for the optimum loading of narrow necked glassware
Brand:  Miele Professional™ 3809340
Code : 04
Additional Details : Weight : 1.00000kg
Product Code. 13172290
  • 1565.00 € / Each
Estimated Shipment: 29-07-2024
to see stock.
Add to basket
Description
Description
  • Equipped with 39 injector nozzles
Specifications
Specifications
Mobile Injector Unit
Narrow-Necked Glassware
1 Each
E 329
4/5 jets 2.5 x 90/110mm, 5/5/5 jets 4.0 x 140/160/180mm, 5/5/5 jets 6.0 x 200/220/240mm, 1 jet for flushing powder dispenser