missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Mobile safety steps, standard duty, anti-slip treads, 5 step
Description
Anti-Slip steps for secure footholds and twin hand rails for user safety. Retractable front castors - hand lever operated. Welded tubular steel with tough nylon castors for easy positioning. 915mm high guard rail to top platform.
- Maximum load capacity: 150kg
- Platform depth: 390
- Number of steps: 5
- Platform height: 1145mm
- Dimensions (H x W x D): 2010mm x 760mm x 1260mm
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction