missing translation for 'onlineSavingsMsg'
Learn More

Harvard Apparatus Mod 1025 Syr 25 ml Tll Terminus

Product Code. 18870771
Change view
Click to view available options
Connection Type:
Luer Lock
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connection Type Unit Size
18870771 Luer Lock Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18870771 Supplier Harvard Apparatus Supplier No. 721835
  • ONLINE EXCLUSIVE PRICE
    436.37 €
    368.24 € / Pack of 1
    Save 68.13 €
    15% Off
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Suitable for dispensing liquids and gases, and high-precision, laboratory/analytical instrument settings

  • 25 ml 1000 Series Gastight™ Syringe
  • PTFE Luer Lock (TLL) Termination
  • PTFE Tipped Plunger (Please see accessory 55-8000 for withdraw/refill applications.)
  • Used dispensing liquids and gases, and in high-precision, laboratory/analytical instrument settings
  • Its 25 mL volume and gastight nature, it may be used for larger volume precision tasks (e.g., reagent delivery, calibration of pumps, instrument priming)

Specifications

Autoclavable Not Autoclavable
Material Glass (Barrel), PTFE (Plunger)
Needle Gauge No Needle Included
Latex Free Yes
Disposable Reusable
Volume (Metric) 25 mL
Connection Type Luer Lock
Graduated Yes
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.