missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Modafinil

Product Code. 18762614
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18762614 10 mg 10mg
18762544 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18762614 Supplier Tocris Bioscience™ Supplier No. 1811/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Psychostimulant

Modafinil is a psychostimulant that displays neuroprotective and antiparkinsonian activity in a primate model of Parkinson's disease. Modafinil promotes vigilance and wakefulness without the central and peripheral side effects associated with conventional dopaminergic psychostimulants. Mechanism of action is not fully understood, possibly via modulation of catecholamine/GABAergic neurotransmission.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(Diphenylmethyl)sulfinyl]acetamide
CAS 68693-11-8
Quantity 10 mg
Target Additional Adrenergic Receptor-related Compounds
Molecular Formula C15H15NO2S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.