Learn More
Hamilton™ Gastight Instrument Syringe
With a precision-machined PTFE plunger tip which creates a leak-free seal.
Brand:  Hamilton™ 208641
Code : NEW
Additional Details : Weight : 0.20000kg
Product Code. 15345104
  • 76.00 € / Each
Estimated Shipment: 27-06-2024
to see stock.
Description
Description
  • Tip wipes the interior of the syringe barrel free of sample
  • Useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze
  • Gas sterilizable
Specifications
Specifications
1 mL