Learn More
Ansell Model 192-G01 - AlphaTec™ 3000 Ultrasonically Welded and Taped Coverall
Product Code. 30259545 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Unit Size:
Pack of 15
Pack of 20
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30259545 Medium Pack of 20
30259537 Small Pack of 20
30259527 Large Pack of 20
30259539 X-Large Pack of 20
30259533 2X-Large Pack of 15
30259543 3X-Large Pack of 15
30259528 4X-Large Pack of 15
30259522 5X-Large Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 30259545 Supplier Ansell Supplier No. YE30T0019203G01
Estimated Shipment: 24-06-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Durable, comfortable multi-layer chemical barrier, Type 3/4/5/6 protection.
  • Multi-layer barrier fabric provides reliable protection against a wide range of chemicals.
  • Innovative safety features, including a dual-zip system, enhance protection.
  • Lightweight yet durable for comfortable wear.
  • Bright yellow high-visibility color helps improve worker safety.
  • Anti-static and tested in accordance with EN 1149-5.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls
Size Medium
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Single-use
Waist Style Elastic
Ankle Style Ties
Cuff Style Locking Glove Ring
Neck Style Hooded
Hood Type 2-panel, Elastic
Closure Type Double Zipper
Seam Type Welded, Taped
Sterility Non-sterile
Resistance Anti-static, Bloodborne Pathogens, Chemicals, Liquid, Particle