Learn More
Jenway™ Model 4510 and Model 4520 Benchtop Conductivity Meter
Description
- Always measure at the best resolution with automatic standard recognition
- Easily calibrate with either standard solution or known cell constant
Specifications
Specifications
| Calibration Points (Conductivity) | 1413 μS, 12.88 mS, 84 μS, 10 μS, 1382 ppm TDS |
| Cell Constant | K = 1 |
| Data Points | Conductivity, TDS, Resistivity, Salinity, Temperature |
| Range (Conductivity) | 0 to 19.99 μS / 0 to 199.9 μS / 0 to 1999 μS / 0 to 19.99 mS / 0 to 199.9 mS / 0 to 1999 mS / 0 to 19.99 S*(only with cell constant >5) |
| Includes | Glass conductivity probe with ATC (K = 1) as noted, electrode holder, and GLP |
| Languages | English, French, German, Italian, Spanish, Portuguese |
| Probe Type | Conductivity Probe |
| Resolution (Conductivity) | 0.01 μS / 0.1 μS / 1 μS / 0.01 mS / 0.1 mS / 1 mS / 0.01 S |