missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Hooded coverall with elastic face opening and adhesive chin flap for tight fit around respirator
- Stitched and over-taped seams
- Double zipper with self-adhesive flap
- Elastic cuffs, ankles and waist,
- Includes thumb loops, integrated socks and boot flaps
- 100% particle tight fabric
- Excellent barrier against a broad range of concentrated inorganic chemicals and biological hazards
- Antistatic treatment (inside)
Specifications
Specifications
| Product Type | Hooded Coveralls |
| Size | Medium |
| Material | Tychem |
| Color | Yellow |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Hood Type | Elastic |
| Closure Type | Double Zipper with Self-adhesive Flap |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?