missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Modifying Enzymes

Product Code. 31722726 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
DNase I
T4 RNA Ligase
T4 RNA Ligase 2
Vaccinia Capping Enzyme
Vaccinia mRNA Cap 2'-O-Methyltransferase
Quantity:
1000 U
1500 U
20,000 U
4000 U
5000 U
Concentration:
10 U/μL
30 U/μL
50 U/μL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Concentration unitSize
31722726 Vaccinia Capping Enzyme 4000 U 10 U/μL Each
31722880 Vaccinia mRNA Cap 2'-O-Methyltransferase 20,000 U 50 U/μL Each
31722960 T4 RNA Ligase 1500 U 30 U/μL Each
31722724 T4 RNA Ligase 2 1000 U 10 U/μL Each
31722812 DNase I 5000 U 50 U/μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31722726 Supplier Thermo Scientific™ Supplier No. EO166AF
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
For Use With (Application) mRNA Capping
Content And Storage 10 U/μL Vaccinia Capping Enzyme, 400 μL

Store at -20°C.
Compatible Buffer 10X Capping Buffer
Quantity 4000 U
Product Type Vaccinia Capping Enzyme
Research Category RNA Research
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.