Learn More
Leica Microsystems Ergo Intermediate Module, DM500/750
Product Code. 12837975
Click to view available options
For Use With (Equipment):
DM500/750 Microscopes
Product Code. 12837975
Brand: Leica Microsystems 13613565
This item requires a price update prior to being ordered.
Request a Price for this item here.
Improve ergonomics by placing this module between the viewing head and the microscope body. Leica Microsystems™ Ergo Intermediate Module, DM500/750 is designed for use with innovative educational microscopes DM500 and DM700.
  • Improves ergonomics.
  • Suitable for DM750 Microscope.
Specifications
Product Type Intermediate Module
For Use With (Equipment) DM500/750 Microscopes