Learn More
Moldex™ Rockets™ Reusable Ear Plugs
Product Code. 12374759
Change view
Click to view available options
Corded:
Uncorded
Plug Color:
Purple
Cord Material:
N/A
Packaging Type:
Pocket-Pak
Unit Size:
Pack of 50
Product Code. Corded Plug Color Cord Material Packaging Type Unit Size
12374759 Uncorded Purple N/A Pocket-Pak Pack of 50
1 options
Product Code. 12374759 Supplier Moldex™ Supplier No. 640001
  • ONLINE EXCLUSIVE PRICE
    234.89 €
    94.22 € / Pack of 50
    Save 140.67 €
    59% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Rockets™ reusable ear plugs are designed to give a secure grip when handling so they easily slip into the ear. Triple-flange design provides maximum comfort for all-day wear.
  • Easy grip handle
  • Air bubble in tip provides cushioned comfort
  • Can be washed and reused
  • 100% PVC-Free™
Specifications
ANSI S3.1974 Yes
Cord Material N/A
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Construction, Energy, Utilities, Manufacturing, Healthcare, Mining, Oil and Gas, Transportation, and Other Various Industries
High Visibility No
Packaging Type Pocket-Pak
Pairs per Package 50 Pairs/Pk.
Plug Material TPE
Plug Color Purple
Noise Reduction Rating (NRR) 27 dB
Corded Uncorded
Plug Shape Flanged
Product Type Reusable Earplugs
Disposable Reusable
Metal Detectable No
Includes Pocket-size case