Learn More
Erlab™ Molecode A
Product Code. 15514520 Shop All Erlab Products
Change view
Click to view available options
For Use With (Equipment):
Captair Smart Fume Hoods
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15514520 Captair Smart Fume Hoods Each
1 options
Product Code. 15514520 Supplier Erlab™ Supplier No. I0699000002
  • ONLINE EXCLUSIVE PRICE
    885.00 €
    866.00 € / Pack of 1
    Save 19.00 €
    2% Off
 Restricted Item
Estimated Shipment: 04-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Erlab™ Molecode A is a saturation detection alarm for broad spectrum filters.
  • Equipment required by class I of the AFNOR standard NF X 15-211: 2009
  • A cell located in the sampling chamber allows the detection automatic saturation of the filter by acids
  • A second cell is placed in contact with the environment of the laboratory indicates a possible pollution of the ambient air
Specifications
For Use With (Equipment) Captair Smart Fume Hoods
Product Type Saturation Detection Alarm
For Use With (Application) Detecting Acids