Learn More
Erlab™ Molecode F
Description
- Equipment required by class I of the AFNOR NF X 15-211: 2009 standard
- A cell located in the sampling chamber allows detection Automatic saturation of the filter by formaldehyde
- A second cell is placed in contact with the environment of the laboratory indicates a possible pollution of the ambient air
Specifications
Specifications
| For Use With (Equipment) | Captair Smart Fume Hoods |
| Product Type | Saturation Detection Alarm |
| For Use With (Application) | Detecting Formaldehyde |