missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ A-23187 Free Acid (Calcimycin)

Product Code. 11524007
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11524007 10 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11524007 Supplier Invitrogen™ Supplier No. A1493
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Commonly used for in situ calibrations of fluorescent Ca2+ indicators

  • Typically used to equilibrate intracellular and extracellular Ca2+ concentrations
  • To permit Mn2+ to enter cell to quench intracellular dye fluorescence
  • Although the intrinsic fluorescence of A-23197 is too high for use with UV-excitable calcium indicators like fura-2, indo-1 and quin-2 it is suitable for use with visible light-excitable indicators, including fluo-4, fluo-3, Calcium Green and rhod-2
  • Can also be used to equilibrate extracelular and intrcellular Mg2+ concentrations, making it useful for calibrating Mg2+ indicators
  • Sustainable packaging

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer, Fluorometer, Fluorescent Imager
Indicator Calcium Indicator
Detection Method Fluorescence
Molecular Formula C29H37N3O6
Physical Form Solid
Product Type Calcium Ionophore

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.