missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Acridine Orange (10 mg/mL in water)

Product Code. 11504047
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11504047 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11504047 Supplier Invitrogen™ Supplier No. A3568
Spotlight Offers
Use promo code 28037
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cell-permeant nucleic acid binding dye

Acridine Orange emits green fluorescence when bound to dsDNA and red fluorescence when bound to ssDNA or RNA. This unique characteristic makes acridine orange useful for cell-cycle studies. Acridine orange has also been used as a lysosomal dye. This dye is available in solution (A-3568).

Apoptosis, Cell Analysis, Cell Cycle, Cell Structure, Cell Tracing & Tracking, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, DNA Fragmentation, Nucleus, Nucleoli & Nuclear Envelope, Organelle Tracing

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Excitation Wavelength Range 460/650 (RNA)
Dye Type Cell-Permeant
For Use With (Equipment) Fluorescence Microscope
Quantity 10 mL
Detection Method Fluorescence
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Dye
Sub Cellular Localization Nucleic Acids, Nucleus
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.