missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Acridine Orange

Product Code. 10553053
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10553053 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10553053 Supplier Invitrogen™ Supplier No. A1301
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Cell-permeant nucleic acid binding dye

Acridine Orange emits green fluorescence when bound to dsDNA and red fluorescence when bound to ssDNA or RNA. This unique characteristic makes acridine orange useful for cell-cycle studies. Acridine orange has also been used as a lysosomal dye. This dye is available in solution (A-3568).

Apoptosis, Cell Analysis, Cell Cycle, Cell Structure, Cell Tracing & Tracking, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, DNA Fragmentation, Nucleus, Nucleoli & Nuclear Envelope, Organelle Tracing

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Content And Storage Store at room temperature and protect from light.
Excitation Wavelength Range 500⁄526 (DNA), 460⁄650 (RNA)
Dye Type Cell-Permeant
For Use With (Equipment) Fluorescence Microscope
Quantity 1 g
Detection Method Fluorescence
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Dye
Sub Cellular Localization Nucleic Acids, Nucleus
Show More Show Less
compliance-icons
Product Identifier
  • acridine orange
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: acridine orange

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.