missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Albumin from Bovine Serum (BSA), Alexa Fluor™ 680 conjugate

Product Code. 11578936
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11578936 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11578936 Supplier Invitrogen™ Supplier No. A34787
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Suited for applications that require well-defined molecular weight tracers

This albumin from bovine serum (BSA), Alexa Fluor 680 conjugate is a bright and photostable conjugate for applications that include studying the rate of endocytosis and exocytosis and following intracellular protein processing.

Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Conjugate Alexa Fluor 680
Species Bovine
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Albumin Proteins
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.