missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 488 Reactive Dye Decapack, for microarrays

Product Code. 10206402
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10206402 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10206402 Supplier Invitrogen™ Supplier No. A32750
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Matches the imaging channels of most popular microarray scanners

The Alexa Fluor™ reactive dye decapack is specially packaged succinimidyl ester dye for labeling of DNA or RNA samples that have been modified by incorporation of aminoallyl-dUTP (Cat. No. A21664) or aminohexylacrylamido-dUTP (Cat. No. A32761). Every packaging lot is rigorously tested for the ability to label aminoallyl-modified DNA to high and consistent levels.

Array CGH, DNA and RNA Purification and Analysis, DNA Labeling, Expression Array Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Microarray Analysis, Nucleic Acid Labeling and Oligo Synthesis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Reactive Dye
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label or Dye Alexa Fluor Dye
Labeling Method Indirect Labeling
Shipping Condition Room Temperature
Product Line Alexa Fluor
Quantity 1 Set

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.